ContactUsLinkComponent
Silicone-free laboratory grease, glisseal N
Silicone-free laboratory grease, glisseal N
Catalog # 291-1111
Supplier:  Borer Chemie
Silicone-free laboratory grease, glisseal N
Catalog # 291-1111
Supplier:  Borer Chemie
Specifications
  • Description:
    glisseal N, normal quality
  • Seq:
    03
  • Environmentally Preferable:
  • Pk:
    1 kg
Specifications
About this item
Silicone-free laboratory grease for sealing and lubricating fixed and rotary ground glass joints and for greasing fittings made of plastics and metall. Withstands vacuum conditions.
  • Apply only a very thin layer
  • Withstands vacuum conditions
  • Excellent thermal stability, constant lubricating consistency between −40 and +200 °C, in the short run even up to 300 °C
  • No dripping point
  • Chemically stable against most acids, alkalis and gases
  • Silicone-free
  • Can be easily removed with the deconex® cleaning concentrates
  • Technical support from the manufacturer and distributor