Orders LinkContactUsLinkComponent
Sample Cups for Analysers
Sample Cups for Analysers
Supplier:   KARTELL
Sample Cups for Analysers
Supplier:  KARTELL
Certificates
Sample Cups for Analysers
Sample Cups for Analysers
10 Options Available Below
Product Details & Documents
PS, transparent.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 391-1017
    For Centrichem® analysers
    For Centrichem® analysers
    0,25 ml
    16,4 mm
    13,7 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 44,90 €
    44,90 €
    1 * 1.000 ST
       44,90 €
  • 391-1018
    For Gemsaec® analysers
    For Gemsaec® analysers
    0,50 ml
    24,5 mm
    13,55 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 33,40 €
    33,40 €
    1 * 1.000 ST
       33,40 €
  • 391-1016
    For Technicon®, Konelab®, Beckman® analysers
    For Technicon®, Konelab®, Beckman® analysers
    1,5 ml
    22,6 mm
    13,8 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 30,00 €
    30,00 €
    1 * 1.000 ST
       30,00 €
  • KART2503
    For Technicon®, Konelab® analysers
    For Technicon®, Konelab® analysers
    1,5 ml
    24,7 mm
    13,9 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 37,60 €
    37,60 €
    1 * 1.000 ST
       37,60 €
  • 212-6120
    For Technicon®, Konelab® analysers
    For Technicon®, Konelab® analysers
    2,0 ml
    24,9 mm
    13,7 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 36,10 €
    36,10 €
    1 * 1.000 ST
       36,10 €
  • 391-1020
    For Hitachi® analysers
    For Hitachi® analysers
    2,0 ml
    38 mm
    16,75 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 49,40 €
    49,40 €
    1 * 1.000 ST
       49,40 €
  • 391-5631
    For Boehringer ES 300® and ES 600® systems
    For Boehringer ES 300® and ES 600® systems
    2,5 ml
    40 mm
    11 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 27,90 €
    27,90 €
    1 * 500 ST
       27,90 €
  • 212-6126
    For Technicon®, Konelab® analysers
    For Technicon®, Konelab® analysers
    4,0 ml
    37,9 mm
    17,26 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 66,90 €
    66,90 €
    1 * 1.000 ST
       66,90 €
Caps
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 391-1019
    Special cross cut cap for Centrifichem® and Gemsaec® analyser cups
    Special cross cut cap for Centrifichem® and Gemsaec® analyser cups
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 33,60 €
    33,60 €
    1 * 1.000 ST
       33,60 €
  • 216-1742
    Sample cup stoppers
    Sample cup stoppers
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 43,10 €
    43,10 €
    1 * 1.000 ST
       43,10 €