Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample tubes

Sample tubes

Supplier:   Simport Scientific
Sample tubes
Supplier:  Simport Scientific
Certificates
Sample tubes

2 Options Available Below

Product Details & Documents

These tubes are chemically resistant and shatterproof, they are supplied with a leakproof screw cap, particularly important when transporting hazardous material.

  • Tube made of PP
  • Cap made of PE
  • Self-standing base with graduation
  • Leakproof screw cap
  • Available sterile and non-sterile
  • Functional temperature range: –90 to +121 °C

Conical bottom tubes with self-standing base graduated from 10 to 30 ml and 10 to 50 ml respectively in 5 ml increments. Ideal for transport and storage of urine, sputum and most liquids or particulate samples. Molded-in graduations make them easy to read.

Packaging: Please note that the non-sterile caps and tubes are package separately in packages of 100. Sterile tubes are supplied in packs of 25.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904

Product Family Options

Product Information

  • Capacity
    Ext.Ø
    Height
    Cap colour
    Version
    Availability
    Price
  • 30 ml
    25,3 mm
    111 mm
    White
    sterile
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 261,00 €
    261,00 €
    1 * 500 ST
       261,00 €

  • 30 ml
    25,3 mm
    111 mm
    White
    non sterile
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 224,00 €
    224,00 €
    1 * 500 ST
       224,00 €