Systainer transport cases
Catalog # 216-4711
Supplier: TRANSPOSAFE
Specifications
- Pk:1
Specifications
About this item
Can be linked, UN approved, for internal and external transport of diagnostic specimens.
- Lid for document holder
- Several Systainers can be linked together with a click system
- Further secured with straps
In a type 3 or 4 Systainer, several small or large safety bags can be sent. The Systainer has a levelling mat at the bottom on which an absorbent sheet of filtration cardboard is placed to absorb any liquid that may leak during transport.