Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Vessel for pathology dispatch
Vessel for pathology dispatch
Supplier:   BOETTGER PAUL
Vessel for pathology dispatch
Supplier:  BOETTGER PAUL
Certificates
Vessel for pathology dispatch
Vessel for pathology dispatch
3 Options Available Below
Product Details & Documents
Polypropylene, extremely clear with HDPE screw cap.
  • Tight fitting screw cap
  • Stable base
  • 35 ml vessel used to protect the 20 ml vessel, 100 ml vessel used to protect the 50 ml vessel
Specially rounded base to facilitate removal of small tissue samples.
Product Family Options
Product Information
  • Capacité
    Ø ext.
    Hauteur
    Couleur bouchon
    Availability
    Price
  • 20 ml
    31 mm
    48 mm
    bleu
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 179,00 €
    179,00 €
    1 * 1.000 ST
       179,00 €
  • 35 ml
    39 mm
    55 mm
    bleu
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 115,00 €
    115,00 €
    1 * 500 ST
       115,00 €
  • 100 ml
    48 mm
    80 mm
    bleu
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 103,00 €
    103,00 €
    1 * 250 ST
       103,00 €