Order Entry
Belgium
ContactUsLinkComponent
 
Affinity chromatography columns, HisTrap™ FF
 :  Cytiva
 
undefined
Affinity chromatography columns, HisTrap™ FF
 :  Cytiva

 

  • Catalog No:
  • 17-5319-01
  • 17-5255-01
  • Max. pressure:
  • 5 bar
  • 5 bar
  • pH range:
  • 3 - 12
  • 3 - 12
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

 

Frequently Bought Together