Order Entry
Belgium
ContactUsLinkComponent
Iron standard solution, 1000 mg/L Fe in 0.5 M nitric acid standard for AAS
Iron standard solution, 1000 mg/L Fe in 0.5 M nitric acid standard for AAS
Catalog # REGEAAFEH
Supplier:  REAGECON
CAS Number:  
Iron standard solution, 1000 mg/L Fe in 0.5 M nitric acid standard for AAS
Catalog # REGEAAFEH
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Iron
  • Symbol:
    Fe

Specifications

About this item