Dropping bottles, LDPE, Wheaton®
Supplier: DWK Life Sciences
Jump to:
About this item
Dropping bottles made of natural or white LDPE. The white LDPE bottles tends to protect the sensitive samples from UV light.
- Good chemical resistance
- Good temperature resistance
26 Options Available Below
Product Details & Documents
Dropping bottles made of natural or white LDPE. The white LDPE bottles tends to protect the sensitive samples from UV light.
- Good chemical resistance
- Good temperature resistance
Product Family Options
Natural
Product Information
- CapacityHeightExt.ØThreadAvailabilityPrice
White
Product Information
- CapacityHeightExt.ØThreadAvailabilityPrice
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.