Decappers, E-Z Decappers™, WHEATON®
These decappers are used to remove aluminium seals from bottles and vials with crimp or serum finish.
- Cushioned ergonomic handle reduces hand fatigue
- Labeled for quick size identification
- Autoclavable
Frequently Bought Together