Orders LinkContactUsLinkComponent
Baskets for test tubes
Baskets for test tubes
Basket for washing and storing test tubes, aluminium, M, 256×152×152 mm
Catalog # HEATHS20341B
Supplier:   HEATHROW
Baskets for test tubes
Catalog # HEATHS20341B
Supplier:  HEATHROW
Baskets for test tubes
Baskets for test tubes
Basket for washing and storing test tubes, aluminium, M, 256×152×152 mm
Price
60,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
These test tube baskets are made of aluminium.
  • Lightweight
  • Autoclavable
  • Sturdy
They are designed for rinsing, transporting and storing laboratory glassware.
Specifications
  • Description:
    Basket for washing and storing test tubes, aluminium, M
  • W×D×H:
    256×152×152 mm