Orders LinkContactUsLinkComponent
Sample spoons, mini, SP Bel-Art
Sample spoons, mini, SP Bel-Art
Certificates
Sample spoons, mini, SP Bel-Art
Sample spoons, mini, SP Bel-Art
5 Options Available Below
Product Details & Documents
PS, white.
  • Disposable, made from white high-impact PS
  • Ideal for collecting specific small volumes of powders and other dry materials
  • 0,10 / 0,50 / 1,0 ml models have a rounded bowl for easier emptying of the spoon and a thinner neck that minimises extra material collection
Product Family Options
Product Information
  • Capacity
    Material
    Availability
    Price
  • 0,05 ml
    PS
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 56,70 €
    56,70 €
    1 * 25 ST
       56,70 €
  • 0,10 ml
    PS
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 53,40 €
    53,40 €
    1 * 25 ST
       53,40 €
  • 0,25 ml
    PS
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 57,50 €
    57,50 €
    1 * 25 ST
       57,50 €
  • 0,50 ml
    PS
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 52,80 €
    52,80 €
    1 * 25 ST
       52,80 €
  • 1,0 ml
    PS
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 60,90 €
    60,90 €
    1 * 25 ST
       60,90 €