ContactUsLinkComponent
Syringes for HPLC autosamplers, Hitachi instruments
Syringes for HPLC autosamplers, Hitachi instruments
Catalog # 548-1655
Syringes for HPLC autosamplers, Hitachi instruments
Catalog # 548-1655
Specifications
  • For:
    L7200/7250, AS1000/2000/4000 LC
  • Type:
    500C-HITACHI
  • Volume:
    500 µl
  • Description:
    Gas-tight, with M10 - 1,0 screw thread termination
  • Pk:
    1