ContactUsLinkComponent
Magnetic stirring bars, PTFE, Spinfin®, SP Bel-Art
Magnetic stirring bars, PTFE, Spinfin®, SP Bel-Art
Catalog # 442-9360
undefined
Magnetic stirring bars, PTFE, Spinfin®, SP Bel-Art
Catalog # 442-9360
Specifications
  • Ø:
    7,9 mm
  • Height:
    9,5 mm
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Spinfin® magnetic stirring bars agitate liquids in round bottom vessels such as test tubes, cylinders and flasks where other stirring bars cannot be used. Bars maintain their position in a vessel and will not wobble.