Magnetic stirring bars, PTFE, octagonal, with pivot ring, Spinbar®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
PTFE encapsulated octagonal magnetic stirring bars with integral pivot ring are the most commonly used shape. Their interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars.
Frequently Bought Together