Orders LinkContactUsLinkComponent
Dewar flasks, flat bottom, type FB
Dewar flasks, flat bottom, type FB
Supplier:   KGW
Dewar flasks, flat bottom, type FB
Supplier:  KGW
Certificates
Dewar flasks, flat bottom, type FB
6 Options Available Below
Product Details & Documents
Dewar flasks are made up of borosilicate glass 3,3 and stainless-steel cover. For storage of liquid nitrogen, dry ice and other coolants.
  • Low evaporation rate
  • Easy handling
  • Antimagnetic
  • Protective casing made of stucco aluminium
Area of application: Biotechnology, medicinal technique, laboratory technique, cooling of small samples and can also be used as insulating flask.
Certifications: Conforms with DIN 12492, ISO 3585.
Specifications for Product Family
Specifications
  • Catalog No:
  • 478-4501
  • KGWA1153
  • KGWA1156
  • KGWA1157
  • KGWA1155
  • KGWA1152
  • Model:
  • FB 0 CAL
  • FB 3 CAL
  • FB 12 CAL
  • FB 18 CAL
  • FB 9 CAL
  • FB 1 CAL
  • Capacity:
  • 0,15 L
  • 0,45 L
  • 1,2 L
  • 2,1 L
  • 0,85 L
  • 0,25 L
  • Daily evaporation:
  • 0,25 L
  • 0,35 L
  • 0,7 L
  • 0,9 L
  • 0,55 L
  • 0,35 L
  • Overall height:
  • 170 mm
  • 220 mm
  • 225 mm
  • 275 mm
  • 220 mm
  • 170 mm
  • Weight, empty:
  • 0,3 kg
  • 0,5 kg
  • 1,2 kg
  • 1,8 kg
  • 0,9 kg
  • 0,4 kg
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • 478-4501
    Dewar flask FB 0 CAL
    Dewar flask FB 0 CAL
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 373,00 €
    373,00 €
    1 * 1 ST
       373,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    478-4501
    Model
    FB 0 CAL
    Capacity
    0,15 L
    Daily evaporation
    0,25 L
    Overall height
    170 mm
    Weight, empty
    0,3 kg
  • KGWA1153
    Dewar flask FB 3 CAL
    Dewar flask FB 3 CAL
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 405,00 €
    405,00 €
    1 * 1 ST
       405,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KGWA1153
    Model
    FB 3 CAL
    Capacity
    0,45 L
    Daily evaporation
    0,35 L
    Overall height
    220 mm
    Weight, empty
    0,5 kg
  • KGWA1156
    Dewar flask FB 12 CAL
    Dewar flask FB 12 CAL
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 514,00 €
    514,00 €
    1 * 1 ST
       514,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KGWA1156
    Model
    FB 12 CAL
    Capacity
    1,2 L
    Daily evaporation
    0,7 L
    Overall height
    225 mm
    Weight, empty
    1,2 kg
  • KGWA1157
    Dewar flask FB 18 CAL
    Dewar flask FB 18 CAL
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 537,00 €
    537,00 €
    1 * 1 ST
       537,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KGWA1157
    Model
    FB 18 CAL
    Capacity
    2,1 L
    Daily evaporation
    0,9 L
    Overall height
    275 mm
    Weight, empty
    1,8 kg
  • KGWA1155
    Dewar flask FB 9 CAL
    Dewar flask FB 9 CAL
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 459,00 €
    459,00 €
    1 * 1 ST
       459,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KGWA1155
    Model
    FB 9 CAL
    Capacity
    0,85 L
    Daily evaporation
    0,55 L
    Overall height
    220 mm
    Weight, empty
    0,9 kg
  • KGWA1152
    Dewar flask FB 1 CAL
    Dewar flask FB 1 CAL
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 396,00 €
    396,00 €
    1 * 1 ST
       396,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KGWA1152
    Model
    FB 1 CAL
    Capacity
    0,25 L
    Daily evaporation
    0,35 L
    Overall height
    170 mm
    Weight, empty
    0,4 kg
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Plug for type FB 9 CAL
    Plug for type FB 9 CAL
    KGWA1655
    Log in for availability
    1 * 1 ST - 43,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Plug for type FB 12 CAL
    Plug for type FB 12 CAL
    KGWA1656
    Log in for availability
    1 * 1 ST - 43,90 €