Orders LinkContactUsLinkComponent
Stainless steel manipulator, Corning® HYPERStack®
Stainless steel manipulator, Corning® HYPERStack®
HYPERStack® stainless steel manipulator
Catalog # 734-2491
Supplier:   Corning
Stainless steel manipulator, Corning® HYPERStack®
Catalog # 734-2491
Supplier:  Corning
Stainless steel manipulator, Corning® HYPERStack®
Stainless steel manipulator, Corning® HYPERStack®
HYPERStack® stainless steel manipulator
Price
26 650,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The Corning® HYPERStack™ Stainless Steel Manipulator accommodates both HYPERStack™-36 and CellSTACK-10 vessels.
It handles two units of the same type simultaneously or one unit at a time. It is ergonomically designed and has large, easy to read dials that are clearly marked to identify positions for ease of use in protocol development.
Specifications
  • Description:
    HYPERStack® stainless steel manipulator