Cell culture vessels, Corning® HYPERStack®
About this item
The Corning® HYPERStack® cell culture vessel is a closed system that allows fluid manipulations to occur through preassembled tubing sets. Two ³/₈" ID tubing ports for filling and venting allow filling via bag assembly, sterile bottle connection or pumping. The attached vent filters are 0,2 μm pore, hydrophobic membranes that allow air displacement during filling while minimising the risk of contamination.
- Features 18,000 cm² cell growth area
- Closed system approach with integrally incorporated USP
- Class VI certified C-FLEX tubing for liquid handling manipulations
- Low particulate assembly method acceptable for cell therapy applications
- Corning proprietary gas-permeable film technology
- Corning CellBIND surface treatment for optimal cell attachment and performance
- USP Class VI virgin polystyrene provides excellent optical clarity and mechanical strength
- Gamma sterilized to SAL 10⁻⁶ standard
- 100% leak tested prior to shipping
4 Options Available Below
Product Details & Documents
The Corning® HYPERStack® cell culture vessel is a closed system that allows fluid manipulations to occur through preassembled tubing sets. Two ³/₈" ID tubing ports for filling and venting allow filling via bag assembly, sterile bottle connection or pumping. The attached vent filters are 0,2 μm pore, hydrophobic membranes that allow air displacement during filling while minimising the risk of contamination.
- Features 18,000 cm² cell growth area
- Closed system approach with integrally incorporated USP
- Class VI certified C-FLEX tubing for liquid handling manipulations
- Low particulate assembly method acceptable for cell therapy applications
- Corning proprietary gas-permeable film technology
- Corning CellBIND surface treatment for optimal cell attachment and performance
- USP Class VI virgin polystyrene provides excellent optical clarity and mechanical strength
- Gamma sterilized to SAL 10⁻⁶ standard
- 100% leak tested prior to shipping
Documents
Corning® HYPERStack® Cell Culture Vessels.pdf
Product Literature
Product Family Options
Product Information
- Growth areaClosure typePackedAvailabilityPrice
- 734-3377HYPERStack® - 12 layer cell culture vessel, Corning® CellBIND® surfaceHYPERStack® - 12 layer cell culture vessel, Corning® CellBIND® surface6000 cm²Without cap1 per pack/4 per caseLog in for availability1 * 4 ST3 270,00 €
- 734-3378HYPERStack® - 12 layer cell culture vessel, non-treated surfaceHYPERStack® - 12 layer cell culture vessel, non-treated surface6000 cm²Without cap1 per pack/4 per caseLog in for availability1 * 4 ST3 430,00 €
- 734-3379HYPERStack® - 36 layer cell culture vessel, Corning® CellBIND® surfaceHYPERStack® - 36 layer cell culture vessel, Corning® CellBIND® surface6000 cm²Without cap1 per pack/2 per caseLog in for availability1 * 2 ST3 470,00 €
- 734-3380HYPERStack® - 36 layer cell culture vessel, non-treated surfaceHYPERStack® - 36 layer cell culture vessel, non-treated surface6000 cm²Without cap1 per pack/2 per caseLog in for availability1 * 2 ST3 370,00 €
Accessories
Product Information
- Catalog #Tubing int.ØTubing ext.ØAvailabilityPrice
- Disposable tubing set, ADCF C-flex, sterileDisposable tubing set, ADCF C-flex, sterile734-24879,5 mm12,7 mmLog in for availability1 * 2 ST - 195,00 €
Product Information
- Catalog #AvailabilityPrice
- Roller bottle with easy grip cap, 850 cm², PS, not treated, sterileRoller bottle with easy grip cap, 850 cm², PS, not treated, sterile734-2492Log in for availability1 * 40 ST - 715,00 €
Product Information
- Catalog #Tubing int.ØTubing ext.ØAvailabilityPrice
- Disposable tubing set, MPC, 0,2 μm filter, sterileDisposable tubing set, MPC, 0,2 μm filter, sterile734-24889,5 mm12,7 mmLog in for availability1 * 2 ST - 817,00 €
Product Information
- Catalog #AvailabilityPrice
- HYPERStack® stainless steel manipulator