Nitrogen receivers (schlenk tubes)
Supplier: Lenz Laborglas
Jump to:
About this item
DURAN®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With 14/23 standard ground sockets/cones and PTFE needle-valve stopcock with tube connector.
8 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With 14/23 standard ground sockets/cones and PTFE needle-valve stopcock with tube connector.
Documents
BSE TSE Glass Columns
Certificats BSE/TSE
Product Family Options
With socket
Product Information
With cone