Sulphuric acid 0.005 mol/l (0.01 N) in aqueous solution stabilised for HPLC
Catalog # KRAF18536.3000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:98.08 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2796
ADR:8,II
:Stabilised with benzalkonium chloride
EINECS:231-639-5
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589