
Dipotassium oxalate 10% in aqueous solution, extra pure
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:583-52-8
:
Formula:C₂K₂O₄
MDL Number:MFCD00012464
:166.22 g/mol
Merck Index:12,07820
EINECS:209-506-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together