Curcumin
Glass bottle for solids, 10 g
Catalog # ROTH9469.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
- CAS Number:458-37-7
- Formula:C₂₁H₂₀O₆
- MDL Number:MFCD00008365
- Molecular Weight:368.39 g/mol
- EINECS:207-280-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



