Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Curcumin

Curcumin
Supplier:  Cayman Chemical
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
  • CAS Number:458-37-7
  • Formula:C₂₁H₂₀O₆
  • MDL Number:MFCD00008365
  • Molecular Weight:368.39 g/mol
  • EINECS:207-280-5

 

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 71,00 €
    71,00 €
    1 * 5 g
       71,00 €

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 108,00 €
    108,00 €
    1 * 10 g
       108,00 €

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 355,00 €
    355,00 €
    1 * 50 g
       355,00 €