Curcumin (from Curcuma longa) ≥80% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 239802-100
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:368.39 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:207-280-5
CAS Number:458-37-7
Formula:C₂₁H₂₀O₆
MDL Number:MFCD00008365