Order Entry
Belgium
ContactUsLinkComponent
Mercury standard solution, 20 mg/L Hg in dil. nitric acid with hydrofluoric acid (max. 1%) (from Hg)
Mercury standard solution, 20 mg/L Hg in dil. nitric acid with hydrofluoric acid (max. 1%) (from Hg)
Catalog # CMPLD0BC.20.2NTRF
CAS Number:  
Mercury standard solution, 20 mg/L Hg in dil. nitric acid with hydrofluoric acid (max. 1%) (from Hg)
Catalog # CMPLD0BC.20.2NTRF
CAS Number:  

Specifications

  • Concentration:
    20 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Mercury
  • Symbol:
    Hg

Specifications

About this item