Order Entry
Belgium
ContactUsLinkComponent
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL), Millipore®
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL), Millipore®
Catalog # 581500-500
CAS Number:  
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL), Millipore®
Catalog # 581500-500
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :172.25 g/mol
EINECS:218-760-9
CAS Number:2226-96-2
Formula:C₉H₁₈NO₂
MDL Number:MFCD00006478