Order Entry
Belgium
ContactUsLinkComponent
 
Ammonium-Nitrogen standard solution, 10 mg/L NH₄ (as N) in water
Catalog # WTWO107012
Supplier:  WTW ONLINE
CAS Number:  
Ammonium-Nitrogen standard solution, 10 mg/L NH₄ (as N) in water
Catalog # WTWO107012
Supplier:  WTW ONLINE
CAS Number:  

Specifications

  • Concentration:
    10 ppm
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    Ammonium-Nitrogen standard solution

Specifications

About this item