Order Entry
Belgium
ContactUsLinkComponent
 
Ammonium-Nitrogen standard solution, 1 mg/L NH₄ (as N) in water
  WTWO107010
 :  
Ammonium-Nitrogen standard solution, 1 mg/L NH₄ (as N) in water
  WTWO107010
 :  

 

  • Concentration:
    1 ppm
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    Ammonium-Nitrogen standard solution