Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dextran, M.W. ~ 200,000

Dextran, M.W. ~ 200,000

Marketsource
Supplier:   Serva
Dextran, M.W. ~ 200,000
Supplier:  Serva
Marketsource
Certificates
Dextran, M.W. ~ 200,000

1 Options Available Below

Product Details & Documents
  • CAS Number:9004-54-0
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00130935
  • EINECS:232-677-5
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • 100 g
    Discontinued
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.