Order Entry
Belgium
ContactUsLinkComponent
Bismuth standard solution, 1000 mg/L Bi in 5% nitric acid (from Bi) TraceCERT® standard for ICP, Supelco®
Bismuth standard solution, 1000 mg/L Bi in 5% nitric acid (from Bi) TraceCERT® standard for ICP, Supelco®
Catalog # SUPL05719-100ML
Supplier:  Merck
CAS Number:  
Bismuth standard solution, 1000 mg/L Bi in 5% nitric acid (from Bi) TraceCERT® standard for ICP, Supelco®
Catalog # SUPL05719-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ300
  • Element:
    Bismuth
  • Symbol:
    Bi

Specifications

About this item

Certifications: Traceable to SRM from NIST, tested in ISO 17034 /ISO 17025 accredited laboratory.