Order Entry
Belgium
ContactUsLinkComponent
J.T.Baker®, Bismuth Standard Solution, 10000 mg/L Bi in 10% Nitric Acid (from Bi) Standard for ICP
J.T.Baker®, Bismuth Standard Solution, 10000 mg/L Bi in 10% Nitric Acid (from Bi) Standard for ICP
Catalog # 5721.0100
Supplier:  Avantor
CAS Number:  
J.T.Baker®, Bismuth Standard Solution, 10000 mg/L Bi in 10% Nitric Acid (from Bi) Standard for ICP
Catalog # 5721.0100
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    10% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Bismuth
  • Symbol:
    Bi

Specifications

About this item

Certifications: Traceable to NIST