Order Entry
Belgium
ContactUsLinkComponent
Bismuth standard solution, 10 mg/L Bi in 5% nitric acid (from Bi), standard for ICP, Specpure®
Bismuth standard solution, 10 mg/L Bi in 5% nitric acid (from Bi), standard for ICP, Specpure®
Catalog # 45249.AE
CAS Number:  
Bismuth standard solution, 10 mg/L Bi in 5% nitric acid (from Bi), standard for ICP, Specpure®
Catalog # 45249.AE
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00134033
  • Element:
    Bismuth
  • Symbol:
    Bi

Specifications

About this item

Molecular Formula: Bi in 2% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134033

Certifications: Traceable to NIST