
Sulphuric acid ≥95.75% calibration solution of viscosity acc. to DIN EN ISO 307
KRAF16351.3700
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
:
:98.08 g/mol
:14,08974
:∼335 °C (1013 hPa)
:3 °C
:1.84 g/cm³ (20 °C)
:Ambient
:231-639-5
:1830
:8,II
:GHS05
:7664-93-9
:H₂SO₄
:MFCD00064589