
Sulphuric acid 1.5 mol/l (3 N) in aqueous solution
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
:GHS07
Frequently Bought Together