Lead standard solution, 10000 mg/L Pb in 2 M nitric acid (from Pb) standard for AAS
Catalog # KRAF13198.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:2 M HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Lead
- Symbol:Pb