Order Entry
Belgium
ContactUsLinkComponent
 
Cadmium standard solution, 10 mg/L Cd in 5% nitric acid (from Cd), standard for ICP, Specpure®
  45257.AE
 :  
Cadmium standard solution, 10 mg/L Cd in 5% nitric acid (from Cd), standard for ICP, Specpure®
  45257.AE
 :  

 

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00134050
  • Element:
    Lead
  • Symbol:
    Pb

 

 

Molecular Formula: Pb in 2% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134050

 : Traceable to NIST