Order Entry
Belgium
ContactUsLinkComponent
Arsenic standard solution, 10 mg/L As in 5% nitric acid (from As), standard for ICP, Specpure®
Arsenic standard solution, 10 mg/L As in 5% nitric acid (from As), standard for ICP, Specpure®
Catalog # 45246.AE
CAS Number:  
Arsenic standard solution, 10 mg/L As in 5% nitric acid (from As), standard for ICP, Specpure®
Catalog # 45246.AE
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Environmentally Preferable:
  • MDL No.:
    MFCD00085309
  • Element:
    Arsenic
  • Symbol:
    As

Specifications

About this item

Molecular Formula: As in 2% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085309

Certifications: Traceable to NIST