Arsenic standard solution, 1000 mg/L As in 1 M nitric acid (from As₂O₅) standard for AAS
Catalog # REGEAAAS05H
Supplier: REAGECON
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:1 M HNO₃
- Pack type:Glass bottle
- Pk:500 mL
- Environmentally Preferable:
- Element:Arsenic
- Symbol:As