Order Entry
Belgium
ContactUsLinkComponent
Antimony standard solution, 5000 mg/L Sb in 5 M hydrochloric acid (from SbCl₃) standard for AAS
Antimony standard solution, 5000 mg/L Sb in 5 M hydrochloric acid (from SbCl₃) standard for AAS
Catalog # KRAF03451.2000
Supplier:  Bernd Kraft
CAS Number:  
Antimony standard solution, 5000 mg/L Sb in 5 M hydrochloric acid (from SbCl₃) standard for AAS
Catalog # KRAF03451.2000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Concentration:
    5000 mg/L
  • Matrix:
    5 M HCl
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Antimony
  • Symbol:
    Sb

Specifications

About this item

Certifications: Traceable to NIST.