Order Entry
Belgium
ContactUsLinkComponent
 
Ammonium standard solution, 10000 mg/L NH₄ in water standard for Ion Chromatography
Catalog # 662-0383
Supplier:  WTW
CAS Number:  
Ammonium standard solution, 10000 mg/L NH₄ in water standard for Ion Chromatography
Catalog # 662-0383
Supplier:  WTW
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    H₂O
  • Pk:
    1 L
  • Description:
    Ammonium standard solution

Specifications

About this item