Order Entry
Belgium
ContactUsLinkComponent
Pesticide reference standard, 1,2,3,4-Tetrachlorobenzene 100 µg/ml in isooctane, CIL
Pesticide reference standard, 1,2,3,4-Tetrachlorobenzene 100 µg/ml in isooctane, CIL
Catalog # PROOCILULM61951.2
CAS Number:  
Pesticide reference standard, 1,2,3,4-Tetrachlorobenzene 100 µg/ml in isooctane, CIL
Catalog # PROOCILULM61951.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    634-66-2
  • Description:
    1,2,3,4-Tetrachlorobenzene 100 µg/ml in isooctane
  • Formula:
    C₆H₂Cl₄
  • Description:
    1,2,3,4-Tetrachlorobenzene

Specifications

About this item