Order Entry
Belgium
ContactUsLinkComponent
 
Pyridoxal hydrochloride, off-white powder
 
Pyridoxal hydrochloride, off-white powder

 

  • Identity Test:
    Passes
  • FTIR:
    Conforms to Standard
  • Solubility Test (0.1% aq soln, 15 min.):
    Clear, Colorless Solution C
  • pH (0,1% aq soln):
    2,7 - 3,5
  • UV/VIS l max (water):
    253-5, 290-1, 316-8 nm
  • Melting Point:
    170 - 180°C

 

Frequently Bought Together