Order Entry
Belgium
ContactUsLinkComponent
Pyridoxal hydrochloride
Pyridoxal hydrochloride
Catalog # A0960.0025
Supplier:  PanReac AppliChem
CAS Number:  
Pyridoxal hydrochloride
Catalog # A0960.0025
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :203.62 g/mol
EINECS:200-602-5
CAS Number:65-22-5
Formula:C₈H₁₀ClNO₃
MDL Number:MFCD00012809