Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sulphuric acid
Sulphuric acid 6 mol/l (12 N) in aqueous solution
Sulphuric acid 6 mol/l (12 N) in aqueous solution
Sulphuric acid 6 mol/l (12 N) in aqueous solution
KRAF05761.7020
:
Bernd Kraft
:
Sulphuric acid 6 mol/l (12 N) in aqueous solution
KRAF05761.7020
:
Bernd Kraft
:
Pack type:
Plastic bottle
Pk:
20 L
Seq:
5302
:
:
98.08 g/mol
:
Ambient
:
231-639-5
:
2796
:
8,II
:
GHS05
:
7664-93-9
:
H₂SO₄
:
MFCD00064589