Sulphuric acid 96,00% ≥96.00% calibration solution of viscosity acc. to DIN EN ISO 307
Catalog # KRAF16352.3700
Supplier: Bernd Kraft
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
About this item
:
:98.08 g/mol
Boiling Pt:∼335 °C (1013 hPa)
EINECS:231-639-5
UN:1830
:GHS05
Storage Temperature:Ambient
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589
Melting Pt:10.38
Density:1.84 g/cm³ (20 °C)
ADR:8 II