Galactitol
Glass bottle for solids, 50 mg
Catalog # EDQMG0049000
Supplier: EDQM
Jump to:
Product Details & Documents
Synonyms: Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:608-66-2
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064288
- Molecular Weight:182.17 g/mol
- Storage Temperature:Ambient
- EINECS:210-165-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg



