Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tri-Calcium dicitrate
tri-Calcium dicitrate
tri-Calcium dicitrate
tri-Calcium dicitrate
Catalog # LOHM502041001
Supplier:
LOHMANN
CAS Number:
Expand image
tri-Calcium dicitrate
Catalog # LOHM502041001
Supplier:
LOHMANN
CAS Number:
Specifications
Pk:
5 kg
Seq:
4114
Specifications
About this item
Product Details
:
:
498.44
EINECS:
212-391-7
CAS Number:
813-94-5
Formula:
C₁₂H₁₀Ca₃O₁₄
Product Details