Order Entry
Belgium
ContactUsLinkComponent
Magnesium acetylacetonate, anhydrous ≥98%
Irritant
Magnesium acetylacetonate, anhydrous ≥98%
Catalog # STRE93-1280-25G
CAS Number:  
Irritant
Magnesium acetylacetonate, anhydrous ≥98%
Catalog # STRE93-1280-25G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :222.52 g/mol
Melting Pt: Dec.
EINECS:237-857-7
 :GHS07
CAS Number:14024-56-7
Formula:C₁₀H₁₄MgO₄
MDL Number:MFCD00013497