Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tri-Lithium citrate tetrahydrate
tri-Lithium citrate tetrahydrate
tri-Lithium citrate tetrahydrate
SERA38644.01
:
Serva
:
tri-Lithium citrate tetrahydrate
SERA38644.01
:
Serva
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
281.99 g/mol
:
213-045-8
:
GHS07
:
6080-58-6
:
C₆H₅Li₃O₇·4H₂O
:
MFCD00150030