Order Entry
Belgium
ContactUsLinkComponent
Formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Flammable
Health
Irritant
Formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Certificates
Flammable
Health
Irritant
Formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™

Specification Test Results

  • Assay (GC):
    Min. 99.5 %
  • Assay of formic acid:
    0.093 - 0.107 %
  • Transmittance at 254 nm:
    Min. 90 %
  • Transmittance at 230 nm:
    Min. 15 %
  • Transmittance at 210 nm:
    Min. 5 %
  • Suitability for the LC-MS:
    complying
  • HPLC Gradient at 254 nm:
    Max. 50 mAU
  • Calcium (Ca):
    Max. 0.5 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Magnesium (Mg):
    Max. 0.5 ppm
  • Sodium (Na):
    Max. 2 ppm

Specification Test Results

Related Information

Frequently Bought Together