Order Entry
Belgium
ContactUsLinkComponent
Magnesium chloride hexahydrate
Magnesium chloride hexahydrate
Catalog # RC-068
Supplier:  G-Biosciences
CAS Number:  
Magnesium chloride hexahydrate
Catalog # RC-068
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :203.3 g/mol
Merck Index:13,05684
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:117 °C
Density:1.57 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7791-18-6
Formula:MgCl₂·6H₂O
MDL Number:MFCD00149781